Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDIR1
Entrez Gene ID
824718
Gene Name
RING/U-box superfamily protein
Gene Aliases
AT3G55530, AtSDIR1, SALT- AND DROUGHT-INDUCED RING FINGER1
Chromosome Location
Chr.3: 20595126 - 20597603 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115410.3 NP_191112.1 3 - 4 302 103
GenBank mRNA AF386995.1 - 3 - 4 272 103
AK317055.1 - 3 - 4 289 103
BT006305.1 - 2 - 3 134 103

View Full Product Details