Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRD1
Entrez Gene ID
824861
Gene Name
dicarboxylate diiron protein, putative (Crd1)
Gene Aliases
ACSF, AT3G56940, CHL27, COPPER RESPONSE DEFECT 1, T8M16.1
Chromosome Location
Chr.3: 21076505 - 21078443 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203183.1 NP_001190112.1 2 - 3 487 114
NM_115553.4 NP_191253.1 1 - 2 564 114

View Full Product Details