Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VLN3
Entrez Gene ID
824908
Gene Name
villin 3
Gene Aliases
AT3G57410, ATVLN3, F28O9.1, villin 3
Chromosome Location
Chr.3: 21243136 - 21251159 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339858.1 NP_001319779.1 22 - 23 2558 102
NM_001339860.1 NP_001325645.1 22 - 23 3323 102
NM_001339861.1 NP_001325651.1 22 - 23 3377 102
NM_001339862.1 NP_001325648.1 22 - 23 2992 102
NM_001339863.1 NP_001325647.1 22 - 23 3030 102
NM_001339865.1 NP_001325649.1 21 - 22 2694 102
NM_001339866.1 NP_001325646.1 21 - 22 2732 102
NM_115601.3 NP_567048.1 22 - 23 2596 102
GenBank mRNA AK318626.1 - 21 - 22 2549 102

View Full Product Details