Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VLN3
Entrez Gene ID
824908
Gene Name
villin 3
Gene Aliases
AT3G57410, ATVLN3, F28O9.1, villin 3
Chromosome Location
Chr.3: 21243136 - 21251159 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339861.1 NP_001325651.1 25 - 26 3954 116
NM_001339863.1 NP_001325647.1 25 - 26 3607 116
NM_001339866.1 NP_001325646.1 24 - 25 3309 116
NM_115601.3 NP_567048.1 25 - 26 3173 116
GenBank mRNA AK318626.1 - 24 - 25 3126 116

View Full Product Details