Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PI-4KBETA2
- Entrez Gene ID
-
830794
- Gene Name
- phosphatidylinositol 4-OH kinase beta2
- Gene Aliases
-
AT5G09350, PHOSPHATIDYLINOSITOL 4-OH KINASE BETA2, PI4KBETA2, T5E8.150, T5E8_150, phosphatidylinositol 4-OH kinase beta2
- Chromosome Location
-
Chr.5: 2899001 - 2906160 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120971.3 |
NP_196497.1 |
10 - 11 |
3386 |
74 |
GenBank mRNA |
AK228931.1 |
- |
10 - 11 |
3372 |
74 |
View Full Product Details