Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUVR2
Entrez Gene ID
834422
Gene Name
SET-domain containing protein lysine methyltransferase family protein
Gene Aliases
AT5G43990, MRH10.10, MRH10_10, SDG18, SET DOMAIN PROTEIN 18
Chromosome Location
Chr.5: 17697686 - 17702036 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085233.2 NP_001078702.1 6 - 7 1832 62
NM_001085234.2 NP_001078703.1 6 - 7 1919 62
NM_001203540.1 NP_001190469.1 6 - 7 1955 62
NM_001344513.1 NP_001331768.1 6 - 7 1933 62
NM_001344514.1 NP_001331767.1 6 - 7 1933 62
NM_001344515.1 NP_001331773.1 5 - 6 1991 62
NM_001344516.1 NP_001331766.1 5 - 6 1917 62
NM_001344517.1 NP_001331772.1 6 - 7 1857 62
NM_001344518.1 NP_001331771.1 6 - 7 1843 62
NM_001344519.1 NP_001331770.1 6 - 7 1843 62
NM_001344520.1 NP_001331769.1 6 - 7 1769 62
NM_001344521.1 NP_001331765.1 5 - 6 2081 62
NM_001344522.1 NP_001331764.1 5 - 6 2021 62
NM_001344523.1 NP_001331763.1 5 - 6 2007 62
NM_001344524.1 NP_001331762.1 6 - 7 1993 62
NM_123766.3 NP_568631.1 6 - 7 1845 62
NM_203151.3 NP_974880.1 6 - 7 1886 62

View Full Product Details