Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUVR2
Entrez Gene ID
834422
Gene Name
SET-domain containing protein lysine methyltransferase family protein
Gene Aliases
AT5G43990, MRH10.10, MRH10_10, SDG18, SET DOMAIN PROTEIN 18
Chromosome Location
Chr.5: 17697686 - 17702036 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085233.2 NP_001078702.1 8 - 9 2152 67
NM_001085234.2 NP_001078703.1 8 - 9 2239 67
NM_001203540.1 NP_001190469.1 8 - 9 2275 67
NM_001344513.1 NP_001331768.1 8 - 9 2253 67
NM_001344514.1 NP_001331767.1 8 - 9 2253 67
NM_001344515.1 NP_001331773.1 7 - 8 2311 67
NM_001344516.1 NP_001331766.1 7 - 8 2237 67
NM_001344517.1 NP_001331772.1 8 - 9 2177 67
NM_001344518.1 NP_001331771.1 8 - 9 2163 67
NM_001344519.1 NP_001331770.1 8 - 9 2163 67
NM_001344520.1 NP_001331769.1 8 - 9 2089 67
NM_001344521.1 NP_001331765.1 7 - 8 2401 67
NM_001344522.1 NP_001331764.1 7 - 8 2341 67
NM_001344523.1 NP_001331763.1 7 - 8 2327 67
NM_001344524.1 NP_001331762.1 8 - 9 2313 67
NM_123766.3 NP_568631.1 8 - 9 2165 67
NM_203151.3 NP_974880.1 8 - 9 2206 67

View Full Product Details