The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_124376.4 | NP_568717.2 | 10 - 11 | 1465 | 63 |
NM_203189.2 | NP_974918.1 | 10 - 11 | 1333 | 63 |