The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_124376.4 | NP_568717.2 | 3 - 4 | 618 | 72 |
NM_203189.2 | NP_974918.1 | 3 - 4 | 519 | 72 |