Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERK2
Entrez Gene ID
840320
Gene Name
somatic embryogenesis receptor-like kinase 2
Gene Aliases
AT1G34210, ATSERK2, F23M19.11, F23M19_11, somatic embryogenesis receptor-like kinase 2
Chromosome Location
Chr.1: 12458564 - 12462905 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333091.1 NP_001320467.1 6 - 7 900 104
NM_103144.5 NP_174683.1 5 - 6 1034 104
GenBank mRNA AK229715.1 - 5 - 6 928 104
FJ708645.1 - 5 - 6 520 104

View Full Product Details