Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT1G45110
Entrez Gene ID
841078
Gene Name
Tetrapyrrole (Corrin/Porphyrin) Methylase
Gene Aliases
AT1G45110, F27F5.18, F27F5_18
Chromosome Location
Chr.1: 17052475 - 17054827 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103586.4 NP_175126.2 5 - 6 651 79
GenBank mRNA AK229448.1 - 5 - 6 592 79
BT010558.1 - 5 - 6 554 79

View Full Product Details