Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BZIP61
Entrez Gene ID
824981
Gene Name
Basic-leucine zipper (bZIP) transcription factor family protein
Gene Aliases
AT3G58120, ATBZIP61
Chromosome Location
Chr.3: 21520974 - 21523518 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339908.1 NP_001327593.1 2 - 3 1011 64
NM_115674.4 NP_191371.1 2 - 3 1154 64
GenBank mRNA AF401300.1 - 2 - 3 714 64
AY087356.1 - 2 - 3 934 64
AY093053.1 - 2 - 3 908 64
BT008728.1 - 2 - 3 714 64

View Full Product Details