Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSL11
Entrez Gene ID
825079
Gene Name
glucan synthase-like 11
Gene Aliases
AT3G59100, ATGSL11, glucan synthase-like 11, gsl11
Chromosome Location
Chr.3: 21842964 - 21854117 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115772.4 NP_191469.3 1 - 2 649 95

View Full Product Details