Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAC9
Entrez Gene ID
825146
Gene Name
sacI homology domain-containing protein / WW domain-containing protein
Gene Aliases
ARABIDOPSIS THALIANA SUPPRESSOR OF ACTIN 9, AT3G59770, AtSAC9, SUPPRESSOR OF ACTIN 9
Chromosome Location
Chr.3: 22078972 - 22086236 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035819.1 NP_001030896.1 12 - 13 4851 91
NM_001203209.1 NP_001190138.1 13 - 14 4778 91
NM_001339998.1 NP_001326481.1 13 - 14 4846 91
NM_115839.4 NP_191536.2 13 - 14 4817 91
GenBank mRNA AY227252.1 - 12 - 13 4611 91

View Full Product Details