Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAC9
Entrez Gene ID
825146
Gene Name
sacI homology domain-containing protein / WW domain-containing protein
Gene Aliases
ARABIDOPSIS THALIANA SUPPRESSOR OF ACTIN 9, AT3G59770, AtSAC9, SUPPRESSOR OF ACTIN 9
Chromosome Location
Chr.3: 22078972 - 22086236 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203209.1 NP_001190138.1 3 - 4 732 63
NM_115839.4 NP_191536.2 3 - 4 819 63
GenBank mRNA AY227252.1 - 2 - 3 613 63

View Full Product Details