Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAC9
Entrez Gene ID
825146
Gene Name
sacI homology domain-containing protein / WW domain-containing protein
Gene Aliases
ARABIDOPSIS THALIANA SUPPRESSOR OF ACTIN 9, AT3G59770, AtSAC9, SUPPRESSOR OF ACTIN 9
Chromosome Location
Chr.3: 22078972 - 22086236 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035819.1 NP_001030896.1 7 - 8 2729 98
NM_001203209.1 NP_001190138.1 8 - 9 2656 98
NM_001339998.1 NP_001326481.1 8 - 9 2724 98
NM_115839.4 NP_191536.2 8 - 9 2695 98
GenBank mRNA AY227252.1 - 7 - 8 2489 98

View Full Product Details