Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX11E
Entrez Gene ID
825279
Gene Name
peroxin 11E
Gene Aliases
AT3G61070, peroxin 11E
Chromosome Location
Chr.3: 22604094 - 22606878 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084853.2 NP_001078322.1 5 - 6 525 67
NM_001203220.1 NP_001190149.1 4 - 5 692 67
NM_115971.3 NP_191666.2 4 - 5 614 67
GenBank mRNA BT002962.1 - 4 - 5 583 67
BT004439.1 - 3 - 4 340 67

View Full Product Details