Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG1
Entrez Gene ID
825287
Gene Name
homeodomain GLABROUS 1
Gene Aliases
AT3G61150, HD-GL2-1, HOMEODOMAIN-GLABRA2 1, homeodomain GLABROUS 1
Chromosome Location
Chr.3: 22630331 - 22635155 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115979.5 NP_191674.1 1 - 2 606 141
GenBank mRNA AY050866.1 - 1 - 2 327 141
AY096757.1 - 1 - 2 168 141

View Full Product Details