Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SULTR4;1
Entrez Gene ID
831199
Gene Name
sulfate transporter 4.1
Gene Aliases
AT5G13550, sulfate transporter 4.1
Chromosome Location
Chr.5: 4355276 - 4359692 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121358.3 NP_196859.1 4 - 5 954 129

View Full Product Details