Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEMG2
Entrez Gene ID
831272
Gene Name
Flavin containing amine oxidoreductase family
Gene Aliases
AT5G14220, F18O22.10, F18O22_10, MATERNAL EFFECT EMBRYO ARREST 61, MEE61, PPO2
Chromosome Location
Chr.5: 4582629 - 4587653 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203378.1 NP_001190307.1 8 - 9 553 130
NM_001343324.1 NP_001331496.1 8 - 9 957 130
NM_001343325.1 NP_001331495.1 9 - 10 848 130
NM_001343326.1 NP_001331494.1 9 - 10 659 130
NM_121426.5 NP_196926.2 9 - 10 622 130
GenBank mRNA AY075665.1 - 9 - 10 597 130
AY101523.1 - 9 - 10 564 130

View Full Product Details