Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FDH
Entrez Gene ID
831330
Gene Name
formate dehydrogenase
Gene Aliases
AT5G14780, AtFDH1, FORMATE DEHYDROGENASE, T9L3.80, T9L3_80, formate dehydrogenase
Chromosome Location
Chr.5: 4776733 - 4779526 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121482.4 NP_196982.1 1 - 2 398 97
GenBank mRNA AB023897.1 - 1 - 2 88 97
AF208028.1 - 1 - 2 145 97
AF208029.1 - 1 - 2 137 97
AF217195.1 - 1 - 2 147 97
AY039609.1 - 1 - 2 147 97
AY054285.1 - 1 - 2 143 97
AY081734.1 - 1 - 2 88 97

View Full Product Details