Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNL2
Entrez Gene ID
831354
Gene Name
SIN3-like 2
Gene Aliases
AT5G15020, F2G14.140, F2G14_140, SIN3-like 2
Chromosome Location
Chr.5: 4859127 - 4866034 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203382.2 NP_001190311.1 8 - 9 1450 121
NM_121506.4 NP_197006.2 8 - 9 1442 121

View Full Product Details