Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- COP1
- Entrez Gene ID
-
817857
- Gene Name
- Transducin/WD40 repeat-like superfamily protein
- Gene Aliases
-
ARABIDOPSIS THALIANA CONSTITUTIVE PHOTOMORPHOGENIC 1, AT2G32950, ATCOP1, CONSTITUTIVE PHOTOMORPHOGENIC 1, DEETIOLATED MUTANT 340, DET340, EMB168, EMBRYO DEFECTIVE 168, FUS1, FUSCA 1, T21L14.11, T21L14_11
- Chromosome Location
-
Chr.2: 13977881 - 13983609 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128855.4 |
NP_180854.1 |
2 - 3 |
507 |
90 |
GenBank mRNA |
BT025337.1 |
- |
2 - 3 |
388 |
90 |
View Full Product Details