Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COP1
Entrez Gene ID
817857
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
ARABIDOPSIS THALIANA CONSTITUTIVE PHOTOMORPHOGENIC 1, AT2G32950, ATCOP1, CONSTITUTIVE PHOTOMORPHOGENIC 1, DEETIOLATED MUTANT 340, DET340, EMB168, EMBRYO DEFECTIVE 168, FUS1, FUSCA 1, T21L14.11, T21L14_11
Chromosome Location
Chr.2: 13977881 - 13983609 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336418.1 NP_001325309.1 5 - 6 1178 84
NM_128855.4 NP_180854.1 5 - 6 1181 84
GenBank mRNA BT025337.1 - 5 - 6 1062 84

View Full Product Details