Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SWI3B
- Entrez Gene ID
-
817927
- Gene Name
- switch subunit 3
- Gene Aliases
-
AT2G33610, ATSWI3B, CHB2, CHROMATIN REMODELING COMPLEX SUBUNIT B, F4P9.38, F4P9_38, switch subunit 3
- Chromosome Location
-
Chr.2: 14228972 - 14231408 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128921.6 |
NP_180919.2 |
4 - 5 |
1168 |
83 |
View Full Product Details