Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MBAC1
Entrez Gene ID
817949
Gene Name
Mitochondrial substrate carrier family protein
Gene Aliases
AT2G33820, ATMBAC1, BAC1, T1B8.12, T1B8_12
Chromosome Location
Chr.2: 14306293 - 14308367 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336471.1 NP_001324477.1 3 - 4 664 105
NM_001336472.1 NP_001324476.1 4 - 5 514 105
NM_001336473.1 NP_001324478.1 4 - 5 490 105
NM_128941.3 NP_180938.2 4 - 5 490 105
GenBank mRNA AY186582.1 - 4 - 5 416 105
BT025874.1 - 4 - 5 416 105

View Full Product Details