Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE18
Entrez Gene ID
817970
Gene Name
maternal effect embryo arrest 18
Gene Aliases
AT2G34090, T14G11.21, T14G11_21, maternal effect embryo arrest 18
Chromosome Location
Chr.2: 14396016 - 14399275 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202741.2 NP_001189670.2 8 - 9 997 103
NM_001336484.1 NP_001323468.1 8 - 9 982 103
NM_001336485.1 NP_001323467.1 8 - 9 877 103
NM_128960.3 NP_180956.2 8 - 9 972 103
NM_201866.2 NP_973595.1 8 - 9 972 103
GenBank mRNA BT002072.1 - 8 - 9 901 103
BT015789.1 - 8 - 9 877 103

View Full Product Details