Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LPAT3
- Entrez Gene ID
-
841549
- Gene Name
- lysophosphatidyl acyltransferase 3
- Gene Aliases
-
AT1G51260, F11M15.12, F11M15_12, lysophosphatidyl acyltransferase 3
- Chromosome Location
-
Chr.1: 19003219 - 19005664 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104004.3 |
NP_175537.1 |
3 - 4 |
397 |
81 |
View Full Product Details