Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGLU36
Entrez Gene ID
841574
Gene Name
beta glucosidase 36
Gene Aliases
AT1G51490, F5D21.16, F5D21_16, beta glucosidase 36
Chromosome Location
Chr.1: 19094888 - 19097503 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333491.1 NP_001319196.1 3 - 4 238 88

View Full Product Details