Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GATL7
Entrez Gene ID
825440
Gene Name
galacturonosyltransferase-like 7
Gene Aliases
AT3G62660, galacturonosyltransferase-like 7
Chromosome Location
Chr.3: 23172754 - 23174993 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116131.5 NP_191825.2 1 - 1 1809 157
GenBank mRNA AY072114.1 - 1 - 1 1632 157

View Full Product Details