Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AtATG18a
Entrez Gene ID
825452
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT3G62770, ATG18a, PEROXISOME UNUSUAL POSITIONING 2, PEUP2, autophagy 18a
Chromosome Location
Chr.3: 23218610 - 23221243 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116142.6 NP_567132.4 3 - 4 1319 71
GenBank mRNA AY039545.1 - 3 - 4 1308 71
AY059169.1 - 3 - 4 1186 71

View Full Product Details