Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEK4
Entrez Gene ID
825503
Gene Name
NIMA-related kinase 4
Gene Aliases
AT3G63280, ATNEK4, MAA21.6, NIMA-RELATED KINASE4, NIMA-related kinase 4
Chromosome Location
Chr.3: 23377976 - 23381590 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125427.1 NP_001118899.1 11 - 12 752 91
NM_001340197.1 NP_001326587.1 12 - 13 791 91
NM_001340198.1 NP_001326588.1 11 - 12 791 91
NM_116193.5 NP_191887.2 12 - 13 832 91
GenBank mRNA AY080682.1 - 12 - 13 826 91
AY117234.1 - 10 - 11 615 91

View Full Product Details