Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADNT1
Entrez Gene ID
827899
Gene Name
adenine nucleotide transporter 1
Gene Aliases
AT4G01100, F2N1.16, F2N1_16, adenine nucleotide transporter 1
Chromosome Location
Chr.4: 477141 - 480028 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203721.1 NP_001190650.1 7 - 8 1291 103
NM_001340300.1 NP_001328407.1 7 - 8 1624 103
NM_116340.4 NP_192019.1 7 - 8 1238 103
GenBank mRNA AF360168.1 - 7 - 8 1233 103
AF412085.1 - 7 - 8 1234 103
AM931440.1 - 7 - 8 979 103
AY056343.1 - 7 - 8 979 103

View Full Product Details