Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NGA4
Entrez Gene ID
826750
Gene Name
AP2/B3-like transcriptional factor family protein
Gene Aliases
AT4G01500, F11O4.9, F11O4_9, NGATHA4
Chromosome Location
Chr.4: 639247 - 640976 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116380.6 NP_192059.4 2 - 2 1286 86

View Full Product Details