Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STS
Entrez Gene ID
828186
Gene Name
stachyose synthase
Gene Aliases
AT4G01970, AtSTS, RS4, T7B11.23, T7B11_23, raffinose synthase 4, stachyose synthase
Chromosome Location
Chr.4: 853922 - 857401 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340350.1 NP_001328930.1 1 - 2 1945 65
NM_116428.5 NP_192106.3 1 - 2 1564 65

View Full Product Details