Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEA3
Entrez Gene ID
825822
Gene Name
K+ efflux antiporter 3
Gene Aliases
AT4G04850, ATKEA3, K+ efflux antiporter 3, T4B21.3, T4B21_3
Chromosome Location
Chr.4: 2452664 - 2457767 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203746.1 NP_001190675.1 9 - 10 1494 91
NM_116723.5 NP_567272.4 9 - 10 1275 91

View Full Product Details