Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAPX
Entrez Gene ID
826396
Gene Name
stromal ascorbate peroxidase
Gene Aliases
AT4G08390, STROMAL ASCORBATE PEROXIDASE, T28D5.80, T28D5_80, stromal ascorbate peroxidase
Chromosome Location
Chr.4: 5314871 - 5317963 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084887.2 NP_001078356.2 8 - 9 1091 109
NM_001340582.1 NP_001319883.1 8 - 9 1128 109
NM_116908.3 NP_192579.1 8 - 9 1106 109
NM_202791.3 NP_974520.1 8 - 9 1094 109
GenBank mRNA AY056319.1 - 8 - 9 1032 109
AY114065.1 - 8 - 9 1000 109

View Full Product Details