Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH7B4
Entrez Gene ID
841849
Gene Name
aldehyde dehydrogenase 7B4
Gene Aliases
AT1G54100, F15I1.19, F15I1_19, aldehyde dehydrogenase 7B4
Chromosome Location
Chr.1: 20194712 - 20199193 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104287.5 NP_175812.1 2 - 3 368 73
NM_179476.3 NP_849807.1 1 - 2 254 73
GenBank mRNA AF378873.1 - 2 - 3 226 73
AJ584645.1 - 1 - 2 136 73
AK230363.1 - - 2033 73
AY048242.1 - 1 - 2 251 73
AY091032.1 - 1 - 2 253 73
AY102145.1 - 1 - 2 134 73
AY117345.1 - 1 - 2 134 73

View Full Product Details