Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PBG1
Entrez Gene ID
842098
Gene Name
20S proteasome beta subunit G1
Gene Aliases
20S proteasome beta subunit G1, AT1G56450, F13N6.3, F13N6_3, PROTEASOME BETA SUBUNIT PBG1
Chromosome Location
Chr.1: 21141633 - 21144458 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104523.4 NP_176040.1 2 - 3 409 125
GenBank mRNA AF378880.1 - 2 - 3 148 125
AK221762.1 - 2 - 3 159 125
AY045596.1 - 2 - 3 125 125
AY052739.1 - 1 - 2 72 125

View Full Product Details