Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AT1G57620
Entrez Gene ID
842138
Gene Name
emp24/gp25L/p24 family/GOLD family protein
Gene Aliases
AT1G57620, CYB, CYTOPLASMIC BODIES, T8L23.9, T8L23_9
Chromosome Location
Chr.1: 21342627 - 21344804 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104559.4 NP_176075.1 1 - 2 438 114
GenBank mRNA AK117498.1 - 1 - 2 344 114
AY085744.1 - 1 - 2 311 114
BT005172.1 - 1 - 2 202 114

View Full Product Details