Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP-PRT1
Entrez Gene ID
842175
Gene Name
ATP phosphoribosyl transferase 1
Gene Aliases
AT1G58080, ATATP-PRT1, ATP PHOSPHORIBOSYL TRANSFERASE, ATP phosphoribosyl transferase 1, HISN1A, T15M6.9, T15M6_9
Chromosome Location
Chr.1: 21504314 - 21507679 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104590.4 NP_176105.1 10 - 11 1389 98
GenBank mRNA AB025249.1 - 10 - 11 1144 98
AY093018.1 - 10 - 11 1163 98
AY128941.1 - 10 - 11 1139 98

View Full Product Details