Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP79C2
Entrez Gene ID
842194
Gene Name
cytochrome p450 79c2
Gene Aliases
AT1G58260, CYP79C3P, F19C14.12, F19C14_12, cytochrome p450 79c2
Chromosome Location
Chr.1: 21605752 - 21607995 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104606.2 NP_176122.2 2 - 3 1223 87

View Full Product Details