Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GolS4
Entrez Gene ID
842342
Gene Name
galactinol synthase 4
Gene Aliases
AT1G60470, AtGolS4, F8A5.2, F8A5_2, galactinol synthase 4
Chromosome Location
Chr.1: 22278975 - 22280593 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104734.4 NP_176250.1 2 - 3 788 80
GenBank mRNA BT023412.1 - 2 - 3 674 80
KJ138958.1 - 2 - 3 649 80

View Full Product Details