Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACOS5
Entrez Gene ID
842596
Gene Name
acyl-CoA synthetase 5
Gene Aliases
AT1G62940, F16P17.9, F16P17_9, acyl-CoA synthetase 5
Chromosome Location
Chr.1: 23310532 - 23312795 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104972.3 NP_176482.1 2 - 3 432 78
GenBank mRNA AY250836.1 - 2 - 3 410 78
AY376734.1 - 2 - 3 410 78

View Full Product Details