Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAT5
Entrez Gene ID
826811
Gene Name
bile acid transporter 5
Gene Aliases
AT4G12030, BASS5, BILE ACID:SODIUM SYMPORTER FAMILY PROTEIN 5, F16J13.100, F16J13_100, bile acid transporter 5
Chromosome Location
Chr.4: 7210807 - 7213376 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340768.1 NP_001319909.1 5 - 6 1415 85
NM_001340769.1 NP_001328579.1 4 - 5 1485 85
NM_001340770.1 NP_001328580.1 5 - 6 1407 85
NM_117273.3 NP_567385.1 2 - 3 997 85
NM_202809.3 NP_974538.1 6 - 7 1337 85
GenBank mRNA AY050449.1 - 6 - 7 1186 85
AY097358.1 - 2 - 3 761 85

View Full Product Details