Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHL1
Entrez Gene ID
826816
Gene Name
AT-hook motif nuclear-localized protein 1
Gene Aliases
AT-hook motif nuclear-localized protein 1, AT4G12080, ATAHL1, F16J13.150, F16J13_150
Chromosome Location
Chr.4: 7239200 - 7241675 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117278.4 NP_192945.2 2 - 3 812 92
GenBank mRNA AB284367.1 - 2 - 3 546 92
AY070738.1 - 2 - 3 793 92
AY149932.1 - 2 - 3 546 92

View Full Product Details