Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE23
Entrez Gene ID
818044
Gene Name
FAD-binding Berberine family protein
Gene Aliases
AT2G34790, EDA28, EMBRYO SAC DEVELOPMENT ARREST 28, F19I3.2, F19I3_2, MATERNAL EFFECT EMBRYO ARREST 23
Chromosome Location
Chr.2: 14673840 - 14677398 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129032.5 NP_181025.1 2 - 3 1140 66
GenBank mRNA BT004022.1 - 2 - 3 1012 66
BT005187.1 - 2 - 3 979 66

View Full Product Details