Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GDCH
Entrez Gene ID
818104
Gene Name
glycine decarboxylase complex H
Gene Aliases
AT2G35370, GLYCINE DECARBOXYLASE COMPLEX H PROTEIN, glycine decarboxylase complex H
Chromosome Location
Chr.2: 14891041 - 14892298 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129089.2 NP_181080.1 3 - 4 566 141
GenBank mRNA AK228462.1 - 3 - 4 433 141
AY050446.1 - 3 - 4 462 141
AY058854.1 - 3 - 4 412 141
AY086345.1 - 3 - 4 505 141
AY097349.1 - 3 - 4 368 141
M82921.1 - 3 - 4 389 141

View Full Product Details