Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YDA
Entrez Gene ID
842674
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT1G63700, EMB71, EMBRYO DEFECTIVE 71, F24D7.11, F24D7_11, MAP KINASE KINASE KINASE 4, MAPKKK4, YODA
Chromosome Location
Chr.1: 23624627 - 23630073 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334122.1 NP_001319310.1 3 - 4 1793 77
NM_105047.2 NP_176557.1 2 - 3 1343 77
GenBank mRNA AY357947.1 - 2 - 3 1343 77

View Full Product Details