Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT1G63750
Entrez Gene ID
842679
Gene Name
Disease resistance protein (TIR-NBS-LRR class) family
Gene Aliases
AT1G63750, F24D7.6, F24D7_6
Chromosome Location
Chr.1: 23650840 - 23655453 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084299.1 NP_001077768.1 3 - 4 2391 89
NM_001084300.2 NP_001077769.1 3 - 4 1986 89
NM_105052.5 NP_176562.2 2 - 3 2800 89

View Full Product Details